Assay Details
| Gene Symbol: Dad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138910.3 | NP_620265.1 | 1 - 2 | 247 | 79 | |
| GenBank mRNA | BC061530.1 | - | 1 - 2 | 251 | 6917861 | 79 |
| FQ223332.1 | - | 1 - 2 | 256 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dad1
Genomic Map