Assay Details
| Gene Symbol: Ccdc125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134761.1 | NP_001128233.1 | 9 - 10 | 1081 | 65 | |
| GenBank mRNA | BC166734.1 | - | 5 - 6 | 940 | 7936477 | 65 |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc125
Genomic Map