Assay Details
| Gene Symbol: LOC100362324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002729007.4 | XP_002729053.2 | 25 - 26 | 4086 | 124 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100362324| Gene Symbol: LOC100362324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002729007.4 | XP_002729053.2 | 25 - 26 | 4086 | 124 | |
Set Membership: |
|