Assay Details
| Gene Symbol: LOC100362324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002729007.4 | XP_002729053.2 | 9 - 10 | 1359 | 145 | |
| XM_006231950.2 | XP_006232012.1 | 9 - 10 | 1359 | 145 | ||
More Information
Set Membership: |
|

LOC100362324
Genomic Map