Assay Details
| Gene Symbol: Crhbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139183.2 | NP_631922.1 | 4 - 5 | 669 | 60 | |
| GenBank mRNA | X58023.1 | - | 4 - 5 | 668 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Crhbp
Genomic Map