Assay Details
| Gene Symbol: Zswim6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006231909.2 | XP_006231971.1 | 7 - 8 | 1345 | 67 | |
| XM_008760730.1 | XP_008758952.1 | 4 - 5 | 3805 | 67 | ||
| GenBank mRNA | AY325185.1 | - | 5 - 6 | 761 | 67 | |
More Information
Set Membership: |
|

Zswim6
Genomic Map