Assay Details
| Gene Symbol: Rfesd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108540.1 | NP_001102010.1 | 2 - 3 | 215 | 71 | |
| XM_006231706.2 | XP_006231768.1 | 3 - 4 | 1315 | 71 | ||
| XM_006231708.2 | XP_006231770.1 | 3 - 4 | 744 | 71 | ||
| XM_006231709.2 | XP_006231771.1 | 3 - 4 | 429 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rfesd
Genomic Map