Assay Details
| Gene Symbol: RGD1562952 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008760706.1 | XP_008758928.1 | 17 - 18 | 1844 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1562952| Gene Symbol: RGD1562952 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008760706.1 | XP_008758928.1 | 17 - 18 | 1844 | 90 | |
Set Membership: |
|