Assay Details
| Gene Symbol: Zfp131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100698.1 | NP_001094168.1 | 2 - 3 | 249 | 122 | |
| XM_006231960.2 | XP_006232022.1 | 2 - 3 | 244 | 122 | ||
| XM_006231961.2 | XP_006232023.1 | 3 - 4 | 473 | 122 | ||
| XM_006231962.2 | XP_006232024.1 | 3 - 4 | 299 | 122 | ||
| XM_006231963.2 | XP_006232025.1 | 3 - 4 | 322 | 122 | ||
| XM_006231965.2 | XP_006232027.1 | 2 - 3 | 399 | 122 | ||
| XM_008760772.1 | XP_008758994.1 | 2 - 3 | 676 | 122 | ||
| GenBank mRNA | BC161974.1 | - | 2 - 3 | 249 | 9029363 | 122 |
| FQ227804.1 | - | 2 - 3 | 519 | 122 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006231965.2 transcripts.
Set Membership: |
|

Zfp131
Genomic Map