Assay Details
| Gene Symbol: Paip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108937.1 | NP_001102407.1 | 7 - 8 | 982 | 149 | |
| XM_008760778.1 | XP_008759000.1 | 7 - 8 | 1280 | 149 | ||
| XM_008760779.1 | XP_008759001.1 | 7 - 8 | 1039 | 149 | ||
| XM_008760780.1 | XP_008759002.1 | 7 - 8 | 1261 | 149 | ||
| GenBank mRNA | BC166996.1 | - | 7 - 8 | 853 | 149 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Paip1
Genomic Map