Assay Details
| Gene Symbol: Paip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108937.1 | NP_001102407.1 | 9 - 10 | 1155 | 176 | |
| XM_008760778.1 | XP_008759000.1 | 9 - 10 | 1453 | 176 | ||
| XM_008760779.1 | XP_008759001.1 | 9 - 10 | 1212 | 176 | ||
| XM_008760780.1 | XP_008759002.1 | 9 - 10 | 1434 | 176 | ||
| GenBank mRNA | BC166996.1 | - | 9 - 10 | 1026 | 176 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Paip1
Genomic Map