Assay Details
| Gene Symbol: Hapln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019189.2 | NP_062062.2 | 1 - 2 | 45 | 87 | |
| GenBank mRNA | BC128740.1 | - | 1 - 2 | 45 | 8371477 | 87 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC128740.1, NM_019189.2 transcripts.
Set Membership: |
|

Hapln1
Genomic Map