Assay Details
| Gene Symbol: Chi3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053560.1 | NP_446012.1 | 3 - 4 | 310 | 74 | |
| XM_006249892.2 | XP_006249954.2 | 3 - 4 | 364 | 74 | ||
| XM_008769574.1 | XP_008767796.1 | 3 - 4 | 331 | 74 | ||
| GenBank mRNA | AF062038.1 | - | 1 - 2 | 191 | 74 | |
| BC091365.1 | - | 3 - 4 | 310 | 7319171 | 74 | |
| EF467168.1 | - | 1 - 2 | 191 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Chi3l1
Genomic Map