Assay Details
| Gene Symbol: Pdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012872.1 | NP_037004.1 | 1 - 2 | 53 | 91 | |
| GenBank mRNA | M33528.1 | - | 1 - 2 | 53 | 91 | |
| M33530.1 | - | 1 - 2 | 63 | 91 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the M33528.1, M33530.1, NM_012872.1 transcripts.
Set Membership: |
|

Pdc
Genomic Map