Assay Details
| Gene Symbol: Mgst3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191594.1 | NP_001178523.1 | 4 - 5 | 279 | 53 | |
| XM_006250209.2 | XP_006250271.1 | 4 - 5 | 365 | 53 | ||
| GenBank mRNA | FQ210409.1 | - | 4 - 5 | 320 | 53 | |
| FQ218175.1 | - | 4 - 5 | 318 | 53 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mgst3
Genomic Map