Assay Details
| Gene Symbol: Thsd7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191669.1 | NP_001178598.1 | 13 - 14 | 2960 | 63 | |
| XM_008769435.1 | XP_008767657.1 | 14 - 15 | 7348 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Thsd7b
Genomic Map