Assay Details
| Gene Symbol: Tnn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107189.2 | NP_001100659.2 | 6 - 7 | 1455 | 88 | |
| XM_008769642.1 | XP_008767864.1 | 7 - 8 | 1852 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P218612 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P808236 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248416 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867387 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346002 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722765 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689937 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118898 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259445 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852179 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812577 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672408 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28888 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815356 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834947 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528728 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84433 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621721 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745020 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421981 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522645 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925612 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512230 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690699 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572450 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190444 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36892 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824537 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691404 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722045 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577169 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437975 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956040 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866534 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119264 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517272 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471913 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309436 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489543 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883125 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683266 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370146 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870316 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47634 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650919 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411412 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193661 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14809 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160754 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26144 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24960 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120715 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30790 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9404 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77285 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P218612 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P808236 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P218612 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P808236 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P808236 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P218612 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P218612 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 69165 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 31548 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10769113 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769127 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769122 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769119 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769109 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769112 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769111 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769130 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769123 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769121 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769116 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769124 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769128 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769115 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769108 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769110 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769129 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769125 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769117 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769107 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769126 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769118 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769114 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769120 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10769106 | - | - |
| Rat Genome 230 2.0 Array | 1379331_at | - | - |
| Rat Genome 230 2.0 Array | 1379331_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1300012105.rn.1 | TC1300001730.rn.1 | RTA TC |
| Rat Expression Set 230 | 1379331_at | - | - |
| Rat Expression Set 230 | 1379331_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10769106 | - | - |
| Rat Genome U34 Set | rc_AA893846_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22272212 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1363912 | - | - |

Tnn
Genomic Map