Assay Details
| Gene Symbol: Mbd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115025.1 | NP_001108497.1 | 2 - 3 | 711 | 79 | |
| GenBank mRNA | BC158877.1 | - | 2 - 3 | 711 | 7934371 | 79 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
heart development
aging
response to mechanical stimulus
response to organic cyclic compound
positive regulation of Wnt signaling pathway
response to nutrient levels
response to estradiol
positive regulation of chromatin binding
regulation of cell proliferation
maternal behavior
ATP-dependent chromatin remodeling
cellular protein complex assembly
regulation of DNA methylation
embryonic organ development
cellular response to organic cyclic compound
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
DNA binding
chromatin binding
mRNA binding
methyl-CpG binding
protein domain specific binding
nucleosomal DNA binding
siRNA binding
C2H2 zinc finger domain binding
negative regulation of transcription from RNA polymerase II promoter
heart development
aging
response to mechanical stimulus
response to organic cyclic compound
positive regulation of Wnt signaling pathway
response to nutrient levels
response to estradiol
positive regulation of chromatin binding
regulation of cell proliferation
maternal behavior
ATP-dependent chromatin remodeling
cellular protein complex assembly
regulation of DNA methylation
embryonic organ development
cellular response to organic cyclic compound
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
DNA binding
chromatin binding
mRNA binding
methyl-CpG binding
protein domain specific binding
nucleosomal DNA binding
siRNA binding
C2H2 zinc finger domain binding
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P583487 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832716 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42729 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329595 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362360 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82252 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650909 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126327 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116811 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377378 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573885 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803202 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469511 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624158 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601193 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709085 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469094 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809974 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730385 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325712 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76325 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414414 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863970 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203812 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365496 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912754 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504735 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920087 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883496 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611577 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275151 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337844 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304290 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912866 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283980 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725787 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503112 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96092 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7221 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132635 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87541 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107874 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26931 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P583487 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P583487 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P583487 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P583487 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 2427 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10802570 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10802573 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10802572 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10802571 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10802568 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10802569 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10802567 | - | - |
| Rat Genome 230 2.0 Array | 1388774_at | - | - |
| Rat Genome 230 2.0 Array | 1388774_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1388774_at | - | - |
| Affymetrix HT Rat Focus Array | 1388774_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1800005582.rn.1 | TC1800000779.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1800005583.rn.1 | TC1800000779.rn.1 | RTA TC |
| Rat Expression Set 230 | 1388774_at | - | - |
| Rat Expression Set 230 | 1388774_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10802567 | - | - |
| Rat Genome U34 Set | rc_AA946379_at | - | - |
| Rat Genome U34 Set | rc_AA850378_at | - | - |
| Rat Genome U34 Set | rc_AA850378_g_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20960056 | - | - |

Mbd2
Genomic Map