Assay Details
| Gene Symbol: Zfp608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107378.1 | NP_001100848.1 | 1 - 2 | 362 | 106 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001107378.1 transcripts.
Set Membership: |
|

Zfp608
Genomic Map