Assay Details
| Gene Symbol: Snx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106135.2 | NP_001099605.2 | 1 - 2 | 149 | 68 | |
| XM_006254716.2 | XP_006254778.1 | 1 - 2 | 175 | 68 | ||
| GenBank mRNA | BC166853.1 | - | 1 - 2 | 124 | 9092757 | 68 |
| CO565239.1 | - | 1 - 2 | 149 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Snx2
Genomic Map