Assay Details
| Gene Symbol: Oacyl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106139.1 | NP_001099609.1 | 13 - 14 | 1810 | 68 | |
| XM_006254859.2 | XP_006254921.1 | 11 - 12 | 1579 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Oacyl
Genomic Map