Assay Details
| Gene Symbol: LOC103694868 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773817.1 | XP_008772039.1 | 2 - 3 | 1872 | 158 | |
| Gene Symbol: LOC100910486 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751788.3 | XP_003751836.2 | 2 - 3 | 1872 | 158 | |
| Gene Symbol: Slc27a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106145.1 | NP_001099615.1 | 6 - 7 | 1479 | 158 | |
More Information
Updated Mapping Information
Set Membership: |
|

LOC103694868
Genomic Map