Assay Details
| Gene Symbol: Wbp11l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008763449.1 | XP_008761671.1 | 250 | 111 | ||
| XM_008763450.1 | XP_008761672.1 | 252 | 111 | |||
| Gene Symbol: Wbp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008763375.1 | 2 - 3 | 250 | 111 | ||
| GenBank mRNA | BC087082.1 | - | 2 - 3 | 163 | 7189215 | 111 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Wbp11l1
Genomic Map