Assay Details
| Gene Symbol: LOC500881 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008765532.1 | XP_008763754.1 | 2 | 1740 | 64 | |
| Gene Symbol: Nop2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191785.1 | NP_001178714.1 | 14 - 15 | 1608 | 64 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC500881
Genomic Map