Assay Details
| Gene Symbol: LOC100911959 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006244623.2 | XP_006244685.1 | 5 - 6 | 767 | 134 | |
| XM_006244624.2 | XP_006244686.1 | 5 - 6 | 766 | 134 | ||
| Gene Symbol: Usp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106619.1 | NP_001100089.1 | 11 - 12 | 1424 | 134 | |
| XM_006237338.1 | XP_006237400.1 | 11 - 12 | 1417 | 134 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100911959
Genomic Map