Assay Details
| Gene Symbol: Crebl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015027.1 | NP_001015027.1 | 5 | 119 | ||
| GenBank mRNA | BC091323.1 | - | 5 | 7313598 | 119 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Crebl2
Genomic Map