Assay Details
| Gene Symbol: Rhno1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027366.1 | 1 - 2 | 402 | 78 | ||
| GenBank mRNA | BC079219.1 | - | 1 - 2 | 402 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rhno1
Genomic Map