Assay Details
| Gene Symbol: Usp18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014058.1 | NP_001014080.1 | 1 - 2 | 261 | 85 | |
| GenBank mRNA | BC079101.1 | - | 1 - 2 | 261 | 7124857 | 85 |
More Information
Updated Mapping Information
Set Membership: |
|

Usp18
Genomic Map