Assay Details
| Gene Symbol: Cd163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107887.1 | NP_001101357.1 | 10 - 11 | 2564 | 61 | |
| XM_006237352.2 | XP_006237414.1 | 10 - 11 | 2593 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P053012 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340160 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590348 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925591 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2328 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250739 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317745 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17097 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136789 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292006 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370974 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137068 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146802 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481981 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337419 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572335 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540705 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869158 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177365 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304981 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544207 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648040 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202134 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57086 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22207 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49823 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P053012 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P053012 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P053012 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P17457 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P18105 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 82839 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10858640 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858631 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858627 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858633 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858636 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858637 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858639 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858628 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858629 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858632 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858638 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858630 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858634 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858635 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10858626 | - | - |
| Rat Genome 230 2.0 Array | 1393917_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1379716_at | - | - |
| Rat Genome 230 2.0 Array | 1393917_at | - | - |
| Rat Genome 230 2.0 Array | 1379716_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1393917_at | - | - |
| Affymetrix HT Rat Focus Array | 1393917_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0400012678.rn.1 | TC0400001748.rn.1 | RTA TC |
| Rat Expression Set 230 | 1393917_PM_at | - | - |
| Rat Expression Set 230 | 1379716_PM_at | - | - |
| Rat Expression Set 230 | 1393917_at | - | - |
| Rat Expression Set 230 | 1379716_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858626 | - | - |
| Rat Genome U34 Set | rc_AA901107_at | - | - |
| Rat Genome U34 Set | rc_AA956555_at | - | - |
| Rat Genome U34 Set | rc_AA901107_g_at | - | - |
| Rat Genome U34 Set | rc_AI234710_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22369942 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1350943 | - | - |

Cd163
Genomic Map