Assay Details
| Gene Symbol: Crebl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015027.1 | NP_001015027.1 | 1 - 2 | 164 | 67 | |
| XM_008763397.1 | XP_008761619.1 | 1 - 2 | 312 | 67 | ||
| GenBank mRNA | BC091323.1 | - | 1 - 2 | 164 | 7313598 | 67 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC091323.1, NM_001015027.1, XM_008763397.1 transcripts.
Set Membership: |
|

Crebl2
Genomic Map