Assay Details
| Gene Symbol: Crebl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015027.1 | NP_001015027.1 | 4 - 5 | 532 | 269 | |
| GenBank mRNA | BC091323.1 | - | 4 - 5 | 532 | 7313598 | 269 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
cell differentiation
positive regulation of peptidyl-serine phosphorylation
positive regulation of fat cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of glucose import
positive regulation of lipid biosynthetic process
protein stabilization
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
transcription, DNA-templated
regulation of transcription, DNA-templated
cell differentiation
positive regulation of peptidyl-serine phosphorylation
positive regulation of fat cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of glucose import
positive regulation of lipid biosynthetic process
protein stabilization
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P590239 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P094200 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359837 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912672 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737370 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914388 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881154 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 713761 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441174 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873319 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726285 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748196 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921024 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542802 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494349 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724451 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414739 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600939 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377216 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699241 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527931 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504952 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582566 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17080 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161230 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141526 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134615 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97191 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109925 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P590239 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P094200 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P590239 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P094200 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P094200 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P590239 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P519174 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 2341 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 13892 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 99351 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 45235 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10859251 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10859250 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10859253 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10859252 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10859249 | - | - |
| Rat Genome 230 2.0 Array | 1390399_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1390399_at | - | - |
| Rat Genome 230 2.0 Array | 1373618_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1373618_at | - | - |
| Affymetrix HT Rat Focus Array | 1373618_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1390399_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1390399_at | - | - |
| Affymetrix HT Rat Focus Array | 1373618_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0400013909.rn.1 | TC0400001923.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400013908.rn.1 | TC0400001923.rn.1 | RTA TC |
| Rat Expression Set 230 | 1390399_at | - | - |
| Rat Expression Set 230 | 1390399_PM_at | - | - |
| Rat Expression Set 230 | 1373618_at | - | - |
| Rat Expression Set 230 | 1373618_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10859249 | - | - |
| Rat Genome U34 Set | rc_AA818359_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1650407 | - | - |

Crebl2
Genomic Map