Assay Details
| Gene Symbol: Lpcat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012189.1 | NP_001012189.1 | 7 - 8 | 836 | 74 | |
| GenBank mRNA | BC089869.1 | - | 7 - 8 | 836 | 7324068 | 74 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P17428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845026 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583714 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451600 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821205 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281525 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18051 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328223 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414166 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156892 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740801 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21207 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683290 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313484 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252204 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264104 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253675 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86813 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652549 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102447 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566041 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575786 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399982 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847878 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593621 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598706 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6228 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550364 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258511 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114221 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40560 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46723 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 29014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97379 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150472 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P17428 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P17428 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P17428 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P17428 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 42292 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 61931 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 15409 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 56461 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10858675 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858678 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858681 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858684 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858682 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858674 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858676 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858685 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858683 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858680 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858679 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858673 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858677 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10858672 | - | - |
| Rat Genome 230 2.0 Array | 1393915_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1383622_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1376813_at | - | - |
| Rat Genome 230 2.0 Array | 1393915_at | - | - |
| Rat Genome 230 2.0 Array | 1376813_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1383622_at | - | - |
| Affymetrix HT Rat Focus Array | 1376813_at | - | - |
| Affymetrix HT Rat Focus Array | 1393915_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1376813_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1393915_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0400012747.rn.1 | TC0400001756.rn.1 | RTA TC |
| Rat Expression Set 230 | 1393915_PM_at | - | - |
| Rat Expression Set 230 | 1393915_at | - | - |
| Rat Expression Set 230 | 1376813_at | - | - |
| Rat Expression Set 230 | 1376813_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10858672 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22069418 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376336 | - | - |
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_2040136 | - | - |
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1375375 | - | - |
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_2040134 | - | - |

Lpcat3
Genomic Map