Assay Details
| Gene Symbol: Atf7ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749858.3 | XP_003749906.1 | 11 - 12 | 3139 | 62 | |
| XM_006237573.2 | XP_006237635.1 | 11 - 12 | 3606 | 62 | ||
| XM_006237575.2 | XP_006237637.1 | 11 - 12 | 3437 | 62 | ||
| XM_006237577.2 | XP_006237639.1 | 11 - 12 | 3157 | 62 | ||
| XM_006237578.2 | XP_006237640.1 | 10 - 11 | 3533 | 62 | ||
More Information
Set Membership: |
|

Atf7ip
Genomic Map