Assay Details
| Gene Symbol: Slc2a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017102.2 | NP_058798.2 | 274 | 121 | ||
| XM_006237295.1 | XP_006237357.1 | 502 | 121 | |||
| XM_006237296.2 | XP_006237358.1 | 821 | 121 | |||
| XM_006237297.1 | XP_006237359.1 | 506 | 121 | |||
| XM_006237298.2 | XP_006237360.1 | 784 | 121 | |||
| GenBank mRNA | BC063168.1 | - | 3 - 4 | 401 | 6887820 | 121 |
| D13962.1 | - | 358 | 121 | |||
| U17978.1 | - | 274 | 121 | |||
| Gene Symbol: LOC100909595 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749833.3 | XP_003749881.1 | 3 - 4 | 608 | 121 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Slc2a3
Genomic Map