Assay Details
| Gene Symbol: Spsb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009660.1 | NP_001009660.1 | 1 - 2 | 131 | 150 | |
| GenBank mRNA | BC088189.1 | - | 1 - 2 | 131 | 7371446 | 150 |
| Gene Symbol: LOC100911548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750655.3 | XP_003750703.1 | 1 - 2 | 243 | 150 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC088189.1,NM_001009660.1,XM_003750655.3 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Spsb2
Genomic Map