Assay Details
| Gene Symbol: Spsb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006237336.2 | XP_006237398.1 | 1 - 2 | 616 | 72 | |
| Gene Symbol: LOC100911548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006244625.2 | XP_006244687.1 | 1 - 2 | 612 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006237336.2,XM_006244625.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Spsb2
Genomic Map