Assay Details
| Gene Symbol: Pde7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031080.1 | NP_112342.1 | 10 - 11 | 1189 | 77 | |
| XM_006232188.2 | XP_006232250.1 | 10 - 11 | 1880 | 77 | ||
| XM_008760880.1 | XP_008759102.1 | 9 - 10 | 2015 | 77 | ||
| GenBank mRNA | U77880.1 | - | 9 - 10 | 894 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P1000449 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P160760 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851032 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812918 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365202 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858897 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433373 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642635 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157082 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574642 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408770 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626127 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578913 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776663 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403828 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921131 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962193 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736647 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400901 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277807 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690439 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171761 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555350 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242934 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19931 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646629 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177017 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865112 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283407 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250277 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76836 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105712 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179270 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768002 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912104 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735485 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60011 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559902 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375019 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238278 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380761 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703369 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240746 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154770 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641549 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583174 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374824 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692472 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375006 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93614 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140486 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151477 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30134 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64788 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43312 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136260 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1000449 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P160760 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P160760 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1000449 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P1000449 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P160760 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1000449 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700527771 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 38512 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 91388 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700527771 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10822456 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10822451 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10822449 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10822452 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10822445 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10822454 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10822457 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10822455 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10822459 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10822446 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10822450 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10822448 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10822453 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10822447 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10822444 | - | - |
| Rat Genome 230 2.0 Array | 1371044_at | - | - |
| Rat Genome 230 2.0 Array | 1382761_at | - | - |
| Rat Genome 230 2.0 Array | 1371044_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1382761_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1371044_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1382761_at | - | - |
| Affymetrix HT Rat Focus Array | 1382761_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1371044_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0200022818.rn.1 | TC0200003339.rn.1 | RTA TC |
| Rat Expression Set 230 | 1371044_at | - | - |
| Rat Expression Set 230 | 1371044_PM_at | - | - |
| Rat Expression Set 230 | 1382761_at | - | - |
| Rat Expression Set 230 | 1382761_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10822444 | - | - |
| Rat Genome U34 Set | U77880_at | - | - |
| Rat Genome U34 Set | rc_AA945945_at | - | - |
| Rat Genome U34 Set | rc_AI059555_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353897 | - | - |

Pde7a
Genomic Map