Assay Details
| Gene Symbol: Car1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107660.1 | NP_001101130.1 | 1 - 2 | 57 | 69 | |
| GenBank mRNA | FQ225569.1 | - | 1 - 2 | 61 | 69 | |
| FQ225719.1 | - | 1 - 2 | 51 | 69 | ||
| FQ231425.1 | - | 1 - 2 | 51 | 69 | ||
| FQ233411.1 | - | 1 - 2 | 53 | 69 | ||
| FQ235246.1 | - | 1 - 2 | 61 | 69 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001107660.1 transcripts.
Set Membership: |
|

Car1
Genomic Map