Assay Details
| Gene Symbol: Car1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107660.1 | NP_001101130.1 | 5 - 6 | 532 | 71 | |
| XM_006232111.1 | XP_006232173.1 | 4 - 5 | 486 | 71 | ||
| XM_008760842.1 | XP_008759064.1 | 5 - 6 | 489 | 71 | ||
| GenBank mRNA | BC158888.1 | - | 5 - 6 | 495 | 8373855 | 71 |
| FQ225390.1 | - | 3 - 4 | 403 | 71 | ||
| FQ225569.1 | - | 5 - 6 | 536 | 71 | ||
| FQ225719.1 | - | 5 - 6 | 526 | 72 | ||
| FQ231425.1 | - | 5 - 6 | 526 | 71 | ||
| FQ233411.1 | - | 5 - 6 | 528 | 71 | ||
| FQ235246.1 | - | 5 - 6 | 536 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Car1
Genomic Map