Assay Details
| Gene Symbol: Car13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134993.1 | NP_001128465.1 | 1 - 2 | 92 | 80 | |
| GenBank mRNA | BC169052.1 | - | 1 - 2 | 56 | 7386405 | 80 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC169052.1,NM_001134993.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Car13
Genomic Map