Assay Details
| Gene Symbol: Car13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134993.1 | NP_001128465.1 | 4 - 5 | 499 | 137 | |
| GenBank mRNA | BC169052.1 | - | 4 - 5 | 463 | 7386405 | 137 |
| FQ220871.1 | - | 4 - 5 | 461 | 137 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Car13
Genomic Map