Assay Details
| Gene Symbol: Slc30a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106404.1 | NP_001099874.1 | 13 - 14 | 1909 | 68 | |
| GenBank mRNA | FQ229887.1 | - | 1 - 2 | 131 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc30a5
Genomic Map