Assay Details
| Gene Symbol: Mctp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008760610.1 | XP_008758832.1 | 8 - 9 | 1349 | 85 | |
| XM_008760611.1 | XP_008758833.1 | 7 - 8 | 1310 | 85 | ||
| XM_008760612.1 | XP_008758834.1 | 9 - 10 | 4029 | 85 | ||
| XM_008760613.1 | XP_008758835.1 | 8 - 9 | 3893 | 85 | ||
| XM_008760614.1 | XP_008758836.1 | 8 - 9 | 1345 | 85 | ||
| XM_008760615.1 | XP_008758837.1 | 7 - 8 | 1306 | 85 | ||
| XM_008760616.1 | XP_008758838.1 | 4 - 5 | 3499 | 85 | ||
| XM_008760617.1 | XP_008758839.1 | 4 - 5 | 380 | 85 | ||
| GenBank mRNA | FQ232558.1 | - | 7 - 8 | 929 | 85 | |
More Information
Set Membership: |
|

Mctp1
Genomic Map