Assay Details
| Gene Symbol: Mocs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007633.2 | NP_001007634.1 | 3 - 4 | 398 | 84 | |
| NM_001162413.1 | NP_001155885.1 | 4 - 5 | 520 | 84 | ||
| XM_006231985.2 | XP_006232047.1 | 4 - 5 | 466 | 84 | ||
| XM_006231986.2 | XP_006232048.1 | 3 - 4 | 363 | 84 | ||
| GenBank mRNA | BC079181.1 | - | 4 - 5 | 486 | 7130858 | 84 |
| CO554692.1 | - | 4 - 5 | 520 | 84 | ||
| FM123325.1 | - | 3 - 4 | 352 | 84 | ||
| FQ220090.1 | - | 4 - 5 | 415 | 84 | ||
| FQ220210.1 | - | 3 - 4 | 311 | 84 | ||
| FQ230755.1 | - | 3 - 4 | 321 | 84 | ||
| FQ235152.1 | - | 4 - 5 | 448 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mocs2
Genomic Map