Assay Details
| Gene Symbol: Ccnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC059113.1 | - | 3 - 4 | 432 | 6922392 | 97 |
| FN800625.1 | - | 3 - 4 | 430 | 97 | ||
| FN800627.1 | - | 1 - 2 | 85 | 97 | ||
| L11995.1 | - | 3 - 4 | 424 | 97 | ||
| X60768.1 | - | 3 - 4 | 362 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccnb1
Genomic Map