Assay Details
| Gene Symbol: Ccnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC059113.1 | - | 5 - 6 | 774 | 6922392 | 78 |
| EV765751.1 | - | 1 - 2 | 179 | 78 | ||
| FN800627.1 | - | 3 - 4 | 427 | 78 | ||
| L11995.1 | - | 5 - 6 | 766 | 78 | ||
| X60768.1 | - | 5 - 6 | 704 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccnb1
Genomic Map