Assay Details
| Gene Symbol: Naip5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_226743.6 | XP_226743.4 | 2 - 3 | 217 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_226743.6 transcripts.
Set Membership: |
|

Naip5
Genomic Map