Assay Details
| Gene Symbol: NEWGENE_1310139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008760704.1 | XP_008758926.1 | 15 - 16 | 3165 | 131 | |
| XM_008760705.1 | XP_008758927.1 | 15 - 16 | 3165 | 131 | ||
| Gene Symbol: Mast4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008760713.1 | XP_008758935.1 | 1817 | 131 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

NEWGENE_1310139
Genomic Map