Assay Details
| Gene Symbol: Polk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138516.1 | NP_612525.1 | 11 - 12 | 1549 | 60 | |
| GenBank mRNA | BC166778.1 | - | 11 - 12 | 1549 | 7934797 | 60 |
More Information
Updated Mapping Information
Set Membership: |
|

Polk
Genomic Map