Assay Details
| Gene Symbol: Cenpk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106407.1 | NP_001099877.1 | 3 - 4 | 581 | 73 | |
| XM_006231888.1 | XP_006231950.1 | 3 - 4 | 581 | 73 | ||
| XM_006231889.2 | XP_006231951.1 | 3 - 4 | 302 | 73 | ||
| XM_006231890.2 | XP_006231952.1 | 3 - 4 | 213 | 73 | ||
| XM_008760667.1 | XP_008758889.1 | 3 - 4 | 581 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cenpk
Genomic Map